Přejít k obsahu
Merck
  • Effects of JS-K, a novel anti-cancer nitric oxide prodrug, on gene expression in human hepatoma Hep3B cells.
Effects of JS-K, a novel anti-cancer nitric oxide prodrug, on gene expression in human hepatoma Hep3B cells.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie (2017-01-26)
Ray Dong, Xueqian Wang, Huan Wang, Zhengyun Liu, Jie Liu, Joseph E Saavedra
ANOTACE
JS-K is a novel anticancer nitric oxide (NO) prodrug effective against a variety of cancer cells, including the inhibition of AM-1 hepatoma cell growth in rats. To further evaluate anticancer effects of JS-K, human hepatoma Hep3B cells were treated with JS-K and the compound control JS-43-126 at various concentrations (0-100μM) for 24h, and cytotoxicity was determined by the MTS assay. The compound control JS-43-126 was not cytotoxic to Hep3B cells at concentrations up to 100μM, while the LC
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
JS-K, ≥97%