Přejít k obsahu
Merck
  • Photoreduction of Shewanella oneidensis Extracellular Cytochromes by Organic Chromophores and Dye-Sensitized TiO
Photoreduction of Shewanella oneidensis Extracellular Cytochromes by Organic Chromophores and Dye-Sensitized TiO
Chembiochem : a European journal of chemical biology (2016-09-30)
Emma V Ainsworth, Colin W J Lockwood, Gaye F White, Ee Taek Hwang, Tsubasa Sakai, Manuela A Gross, David J Richardson, Thomas A Clarke, Lars J C Jeuken, Erwin Reisner, Julea N Butt
ANOTACE
The transfer of photoenergized electrons from extracellular photosensitizers across a bacterial cell envelope to drive intracellular chemical transformations represents an attractive way to harness nature's catalytic machinery for solar-assisted chemical synthesis. In Shewanella oneidensis MR-1 (MR-1), trans-outer-membrane electron transfer is performed by the extracellular cytochromes MtrC and OmcA acting together with the outer-membrane-spanning porin⋅cytochrome complex (MtrAB). Here we demonstrate photoreduction of solutions of MtrC, OmcA, and the MtrCAB complex by soluble photosensitizers: namely, eosin Y, fluorescein, proflavine, flavin, and adenine dinucleotide, as well as by riboflavin and flavin mononucleotide, two compounds secreted by MR-1. We show photoreduction of MtrC and OmcA adsorbed on Ru
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Proflavine hemisulfate salt hydrate, powder