Přejít k obsahu
Merck
  • VCP/p97 cooperates with YOD1, UBXD1 and PLAA to drive clearance of ruptured lysosomes by autophagy.
VCP/p97 cooperates with YOD1, UBXD1 and PLAA to drive clearance of ruptured lysosomes by autophagy.
The EMBO journal (2016-11-01)
Chrisovalantis Papadopoulos, Philipp Kirchner, Monika Bug, Daniel Grum, Lisa Koerver, Nina Schulze, Robert Poehler, Alina Dressler, Sven Fengler, Khalid Arhzaouy, Vanda Lux, Michael Ehrmann, Conrad C Weihl, Hemmo Meyer
ANOTACE
Rupture of endosomes and lysosomes is a major cellular stress condition leading to cell death and degeneration. Here, we identified an essential role for the ubiquitin-directed AAA-ATPase, p97, in the clearance of damaged lysosomes by autophagy. Upon damage, p97 translocates to lysosomes and there cooperates with a distinct set of cofactors including UBXD1, PLAA, and the deubiquitinating enzyme YOD1, which we term ELDR components for Endo-Lysosomal Damage Response. Together, they act downstream of K63-linked ubiquitination and p62 recruitment, and selectively remove K48-linked ubiquitin conjugates from a subpopulation of damaged lysosomes to promote autophagosome formation. Lysosomal clearance is also compromised in MEFs harboring a p97 mutation that causes inclusion body myopathy and neurodegeneration, and damaged lysosomes accumulate in affected patient tissue carrying the mutation. Moreover, we show that p97 helps clear late endosomes/lysosomes ruptured by endocytosed tau fibrils. Thus, our data reveal an important mechanism of how p97 maintains lysosomal homeostasis, and implicate the pathway as a modulator of degenerative diseases.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Leu-Leu methyl ester hydrobromide, ≥97% (TLC)