Přejít k obsahu
Merck
A new triphenylethylene compound, Fc-1157a. I. Hormonal effects.
Cancer chemotherapy and pharmacology (1986-01-01)
S Kallio, L Kangas, G Blanco, R Johansson, A Karjalainen, M Perilä, I Pippo, H Sundquist, M Södervall, R Toivola
ANOTACE
The basic pharmacological and biochemical properties of a new antiestrogen, Fc-1157a, are described. Fc-1157a is bound specifically and with high affinity to estrogen receptors. The binding is competitive with estradiol. Fc-1157a treatment induces translocation of estrogen receptors from cytoplasm to nucleus. The turnover rate of nuclear estrogen receptors is markedly lower than with estradiol, but is more rapid than after tamoxifen. Fc-1157a is an almost pure antiestrogen in rat uterus, but has intrinsic estrogenic activity in mouse uterus. In animal experiments Fc-1157a has shown antitumor properties, which are described in the companion paper.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Toremifene citrate salt, ≥98% (HPLC)