Přejít k obsahu
Merck
  • Combination of secretin and fluvastatin ameliorates the polyuria associated with X-linked nephrogenic diabetes insipidus in mice.
Combination of secretin and fluvastatin ameliorates the polyuria associated with X-linked nephrogenic diabetes insipidus in mice.
Kidney international (2014-02-14)
Giuseppe Procino, Serena Milano, Monica Carmosino, Claudia Barbieri, Maria C Nicoletti, Jian H Li, Jürgen Wess, Maria Svelto
ANOTACE
X-linked nephrogenic diabetes insipidus (X-NDI) is a disease caused by inactivating mutations of the vasopressin (AVP) type 2 receptor (V2R) gene. Loss of V2R function prevents plasma membrane expression of the AQP2 water channel in the kidney collecting duct cells and impairs the kidney concentration ability. In an attempt to develop strategies to bypass V2R signaling in X-NDI, we evaluated the effects of secretin and fluvastatin, either alone or in combination, on kidney function in a mouse model of X-NDI. The secretin receptor was found to be functionally expressed in the kidney collecting duct cells. Based on this, X-NDI mice were infused with secretin for 14 days but urinary parameters were not altered by the infusion. Interestingly, secretin significantly increased AQP2 levels in the collecting duct but the protein primarily accumulated in the cytosol. Since we previously reported that fluvastatin treatment increased AQP2 plasma membrane expression in wild-type mice, secretin-infused X-NDI mice received a single injection of fluvastatin. Interestingly, urine production by X-NDI mice treated with secretin plus fluvastatin was reduced by nearly 90% and the urine osmolality was doubled. Immunostaining showed that secretin increased intracellular stores of AQP2 and the addition of fluvastatin promoted AQP2 trafficking to the plasma membrane. Taken together, these findings open new perspectives for the pharmacological treatment of X-NDI.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-SCTR antibody produced in rabbit, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution