Přejít k obsahu
Merck
  • Urine is a novel source of autologous mesenchymal stem cells for patients with epidermolysis bullosa.
Urine is a novel source of autologous mesenchymal stem cells for patients with epidermolysis bullosa.
BMC research notes (2015-12-15)
Markus Schosserer, Rita Reynoso, Verena Wally, Bogdan Jug, Viktoria Kantner, Sylvia Weilner, Ivana Buric, Johannes Grillari, Johann W Bauer, Regina Grillari-Voglauer
ANOTACE
Regenerative medicine is strictly dependent on stem cells as a source for a high diversity of somatic cells. However, the isolation of such from individuals suffering from severe genetic skin blistering diseases like epidermolysis bullosa (EB) is often associated with further organ damage. Stem cells were isolated from 112 urine samples from 21 different healthy donors, as well as from 33 urine samples from 25 donors with EB. The cultivation of these cells was optimized by testing different media formulations and pre-coating of culture vessels with collagen. The identity of cells was confirmed by testing marker expression, differentiation potential and immune-modulatory properties. We provide here an optimized protocol for the reproducible isolation of mesenchymal stem cells from urine, even from small volumes as obtained from patients with EB. Furthermore, we offer a basic characterization of those urine-derived stem cells (USCs) from healthy donors, as well as from patients with EB, and demonstrate their potential to differentiate into chondrocytes, osteoblasts and adipocytes, as well as their immune-modulatory properties. Thus, USCs provide a novel and non-invasive source of stem cells, which might be applied for gene-therapeutic approaches to improve medical conditions of patients with EB.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Monoclonal Anti-CD44 antibody produced in mouse, clone MEM-263, purified immunoglobulin, buffered aqueous solution