- A long-tethered (P-B-P)-pincer ligand: synthesis, complexation, and application to catalytic dehydrogenation of alkanes.
A long-tethered (P-B-P)-pincer ligand: synthesis, complexation, and application to catalytic dehydrogenation of alkanes.
Dalton transactions (Cambridge, England : 2003) (2016-07-08)
Enrique Huang Kwan, Yasushi Jack Kawai, Sei Kamakura, Makoto Yamashita
PMID27383267
ANOTACE
A new long-tethered boron-containing (P-B-P)-pincer ligand 8 has been synthesized. Complexation of 8 with [Ir(coe)
MATERIÁLY
Číslo produktu
Značka
Popis produktu