Přejít k obsahu
Merck
Synthesis and antiviral activity of P1' arylsulfonamide azacyclic urea HIV protease inhibitors.
Bioorganic & medicinal chemistry letters (2004-07-01)
Peggy P Huang, John T Randolph, Larry L Klein, Sudthida Vasavanonda, Tatyana Dekhtyar, Vincent S Stoll, Dale J Kempf
ANOTACE
A series of novel azacyclic urea HIV protease inhibitors bearing a benzenesulfonamide group at P1' were synthesized utilizing a parallel synthesis method. Structural studies of early analogs bound in the enzyme active site were used to design more potent inhibitors. The effects of substituting the P1' benzenesulfonyl group on antiviral activity and protein binding are described.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
3-Bromobenzenesulfonyl chloride, 96%