Přejít k obsahu
Merck
  • Preparation and pharmacological evaluation of novel glycoprotein (Gp) IIb/IIIa antagonists. 1. The selection of naphthalene derivatives.
Preparation and pharmacological evaluation of novel glycoprotein (Gp) IIb/IIIa antagonists. 1. The selection of naphthalene derivatives.
Chemical & pharmaceutical bulletin (2000-04-05)
S Ono, Y Inoue, T Yoshida, A Ashimori, K Kosaka, T Imada, C Fukaya, N Nakamura
ANOTACE
The synthesis and design using molecular modeling techniques for non-peptide, low molecular weight novel fibrinogen receptor (glycoprotein IIb/IIIa: Gp IIb/IIIa) antagonists, is reported. We used a highly potent serine protease inhibitor, Nafamostat, having an amidinonaphthyl unit as the starting compound. The compounds 4-(6-amidino-2-naphthylaminocarbonyl)phenoxyacetic acid (5a) and 4-(6-amidino-2-naphthalenecarboxamido)phenoxyacetic acid (5b) inhibited adenosin-5'-diphospate (ADP)-induced aggregation of human platelet-rich plasma (PRP) with IC50 values of 0.05 and 0.07 microM, respectively, and had lost their ability to inhibit a variety of serine proteases, including thrombin, factor Xa, plasmin and trypsin.