- In Vitro Studies of Chromone-Tetrazoles against Pathogenic Protozoa, Bacteria, and Fungi.
In Vitro Studies of Chromone-Tetrazoles against Pathogenic Protozoa, Bacteria, and Fungi.
Molecules (Basel, Switzerland) (2015-07-18)
Pedro A Cano, Alejandro Islas-Jácome, Ángeles Rangel-Serrano, Fernando Anaya-Velázquez, Felipe Padilla-Vaca, Elías Trujillo-Esquivel, Patricia Ponce-Noyola, Antonio Martínez-Richa, Rocío Gámez-Montaño
PMID26184131
ANOTACE
In vitro studies to fourteen previously synthesized chromone-tetrazoles and four novel fluorine-containing analogs were conducted against pathogenic protozoan (Entamoeba histolytica), pathogenic bacteria (Pseudomonas aeruginosa, and Staphylococcus aureus), and human fungal pathogens (Sporothrix schenckii, Candida albicans, and Candida tropicalis), which have become in a serious health problem, mainly in tropical countries.