Přejít k obsahu
Merck
  • Lysyl oxidase propeptide stimulates osteoblast and osteoclast differentiation and enhances PC3 and DU145 prostate cancer cell effects on bone in vivo.
Lysyl oxidase propeptide stimulates osteoblast and osteoclast differentiation and enhances PC3 and DU145 prostate cancer cell effects on bone in vivo.
Journal of cell communication and signaling (2015-12-03)
Mona Alsulaiman, Manish V Bais, Philip C Trackman
ANOTACE
Lysyl oxidase pro-enzyme is secreted by tumor cells and normal cells as a 50 kDa pro-enzyme into the extracellular environment where it is cleaved into the ~30 kDa mature enzyme (LOX) and 18 kDa pro-peptide (LOX-PP). Extracellular LOX enzyme activity is required for normal collagen and elastin extracellular cross-linking and maturation of the extracellular matrix. Extracellular LOX-PP acts as a tumor suppressor and can re-enter cells from the extracellular environment to induce its effects. The underlying hypothesis is that LOX-PP has the potential to promote bone cell differentiation, while inhibiting cancer cell effects in bone. Here we investigate the effect of LOX-PP on bone marrow cell proliferation and differentiation towards osteoblasts or osteoclasts, and LOX-PP modulation of prostate cancer cell conditioned media-induced alterations of proliferation and differentiation of bone marrow cells in vitro. Effects of overexpression of rLOX-PP in DU145 and PC3 prostate cancer cell lines on bone structure in vivo after intramedullary injections were determined. Data show that prostate cancer cell conditioned media inhibited osteoblast differentiation in bone marrow-derived cells, which was reversed by rLOX-PP treatment. Prostate cancer conditioned media stimulated osteoclast differentiation which was further enhanced by rLOX-PP treatment. rLOX-PP stimulated osteoclast differentiation by inhibiting OPG expression, up-regulating CCN2 expression, and increasing osteoclast fusion. In vivo studies indicate that rLOX-PP expression by PC3 cells implanted into the tibia of mice further enhanced PC3 cell ability to resorb bone, while rLOX-PP expression in DU145 cells resulted in non-significant increases in net bone formation. rLOX-PP enhances both osteoclast and osteoblast differentiation. rLOX-PP may serve to enhance coupling interactions between osteoclasts and osteoblasts helping to maintain a normal bone turnover in health, while contributing to bone abnormalities in disease.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Osteogenesis Quantitation Kit, The Osteogenesis Quantitation Kit provides all the necessary reagents & a protocol to enable quantification of osteogenesis using a standard plate reader.