Přejít k obsahu
Merck
  • N-(2-Hydroxypropyl)methacrylamide Copolymer-Drug Conjugates for Combination Chemotherapy of Acute Myeloid Leukemia.
N-(2-Hydroxypropyl)methacrylamide Copolymer-Drug Conjugates for Combination Chemotherapy of Acute Myeloid Leukemia.
Macromolecular bioscience (2015-07-30)
Rui Zhang, Jiyuan Yang, Yan Zhou, Paul J Shami, Jindřich Kopeček
ANOTACE
There is a need for new treatment strategies of acute myeloid leukemia (AML). In this study, four different drugs, including cytarabine, daunorubicin, GDC-0980, and JS-K, were investigated in vitro for the two-drug combinations treatment of AML. The results revealed that cytarabine and GDC-0980 had the strongest synergism. In addition, cell cycle analysis was conducted to investigate the effect of the different combinations on cell division. For future in vivo application, N-(2-hydroxypropyl)methacrylamide (HPMA) copolymer-cytarabine and GDC-0980 conjugates were synthesized, respectively. In vitro studies demonstrated that both conjugates had potent cytotoxicity and their combination also showed strong synergy, suggesting a potential chemotherapeutic strategy for future AML treatment.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
JS-K, ≥97%