Přejít k obsahu
Merck
  • Comparison and evaluation of RNA quantification methods using viral, prokaryotic, and eukaryotic RNA over a 10(4) concentration range.
Comparison and evaluation of RNA quantification methods using viral, prokaryotic, and eukaryotic RNA over a 10(4) concentration range.
Analytical biochemistry (2009-05-21)
Roman Aranda, Shauna M Dineen, Rhonda L Craig, Richard A Guerrieri, James M Robertson
ANOTACE
Quantification of RNA is essential for various molecular biology studies. In this work, three quantification methods were evaluated: ultraviolet (UV) absorbance, microcapillary electrophoresis (MCE), and fluorescence-based quantification. Viral, bacterial, and eukaryotic RNA were measured in the 500 to 0.05-ng microl(-1) range via an ND-1000 spectrophotometer (UV), Agilent RNA 6000 kits (MCE), and Quant-iT RiboGreen assay (fluorescence). The precision and accuracy of each method were assessed and compared with a concentration derived independently using inductively coupled plasma-optical emission spectroscopy (ICP-OES). Cost, operator time and skill, and required sample volumes were also considered in the evaluation. Results indicate an ideal concentration range for each quantification technique to optimize accuracy and precision. The ND-1000 spectrophotometer exhibits high precision and accurately quantifies a 1-microl sample in the 500 to 5-ng microl(-1) range. The Quant-iT RiboGreen assay demonstrates high precision in the 1 to 0.05-ng microl(-1) range but is limited to lower RNA concentrations and is more costly than the ND-1000 spectrophotometer. The Agilent kits exhibit less precision than the ND-1000 spectrophotometer and Quant-iT RiboGreen assays in the 500 to 0.05-ng microl(-1) range. However, the Agilent kits require 1 microl of sample and can determine the integrity of the RNA, a useful feature for verifying whether the isolation process was successful.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Roche
RNA, MS2, from bacteriophage MS2