Přejít k obsahu
Merck
  • Bufalin, a component in Chansu, inhibits proliferation and invasion of hepatocellular carcinoma cells.
Bufalin, a component in Chansu, inhibits proliferation and invasion of hepatocellular carcinoma cells.
BMC complementary and alternative medicine (2013-07-23)
Dong-Ze Qiu, Zhou-Ji Zhang, Wei-Zhong Wu, Yun-Ke Yang
ANOTACE
Hepatocellular carcinoma (HCC) is a common and aggressive cancer, and the treatment options are limited for patients with advanced HCC. Bufalin, the major digoxin-like component of the traditional Chinese medicine Chansu, exhibits significant anti-tumor activities in many tumor cell lines. In the present study, we investigated the effect of bufalin on the inhibition of an AKT-related signaling pathway, and examined the relationship between regulatory proteins and anti-tumor effects in hepatoma cells. Proliferation, wound healing, transwell-migration/invasion and adhesion assays were performed in HCCLM3 and HepG2 cell lines. The protein levels of pAKT, AKT, pGSK3β, GSK3β, pβ-catenin, β-catenin, E-cadherin, MMP-9, and MMP-2 were measured by western blot analysis. E-Cadherin and β-catenin expression levels were also evaluated by immunofluorescence. Bufalin inhibited hepatoma cell proliferation, migration, invasion and adhesion. In addition, treatment with bufalin significantly decreased the levels of pAKT, pGSK3β, MMP-9, and MMP-2, while increasing the levels of GSK3β and E-cadherin and suppressing the nuclear translocation of β-catenin. Bufalin is a potential anti-HCC therapeutic candidate through its inhibition of the AKT/GSK3β/β-catenin/E-cadherin signaling pathway. Further studies with bufalin are warranted in patients with HCC, especially those with the disease at advanced stages.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Bufalin