Přejít k obsahu
Merck
Pyrazolo[3,4-d]pyrimidines as adenosine antagonists.
Life sciences (1984-05-28)
L P Davies, S C Chow, J H Skerritt, D J Brown, G A Johnston
ANOTACE
A large number of nitrogen heterocycles structurally related to caffeine and theophylline have been tested for activity as adenosine antagonists. Preliminary screening, utilizing displacement of [3H]N6-phenylisopropyladenosine (PIA) binding to rat brain membranes, identified several pyrazolo[3,4-d]pyrimidines with potential antagonist activity. These were then tested for their ability to antagonize adenosine-stimulated adenylate cyclase of guinea-pig slices and to block adenosine receptors which mediate presynaptic inhibition of transmitter release from cholinergic nerves in guinea-pig ileum. Of several compounds found to have antagonist activity, one of these, 4,6-bis-alpha- carbamoylethylthio -1-phenylpyrazolo[3,4-d]pyrimidine ( DJB -KK) was approximately an order of magnitude more potent than theophylline in both tests. GTP greatly reduces the potency of purine agonists, but not antagonists, as inhibitors of [3H] PIA binding; the potency of the pyrazolo[3,4-d]pyrimidine compounds was not altered by GTP. The compounds have no significant activity against [3H]adenosine uptake or on the binding of ligands to muscarinic cholinergic, beta-adrenergic, GABA or L-glutamate receptors.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Benzoylacetonitrile, 99%