Přejít k obsahu
Merck
  • Genetic Variation in Glutamate Carboxypeptidase II and Interaction with Dietary Natural Vitamin C May Predict Risk for Adenomatous Polyp Occurrence.
Genetic Variation in Glutamate Carboxypeptidase II and Interaction with Dietary Natural Vitamin C May Predict Risk for Adenomatous Polyp Occurrence.
Asian Pacific journal of cancer prevention : APJCP (2015-06-02)
Jeong-Hwa Choi, Zoe Yates, Charlotte Martin, Lyndell Boyd, Xiaowei Ng, Virginia Skinner, Ron Wai, Jeongseon Kim, Hae Dong Woo, Martin Veysey, Mark Lucock
ANOTACE
The C1561T variant of the glutamate carboxypeptidase II (GCPII) gene is critical for natural methylfolylpolyglutamte (methylfolate) absorption, and has been associated with perturbations in folate metabolism and disease susceptibility. However, little is known on C1561T-GCPII as a risk factor for colorectal cancer. Therefore, this study examined whether C1561T-GCPII influences folate metabolism and adenomatous polyp occurrence. 164 controls and 38 adenomatous polyp cases were analysed to determine blood folate and plasma homocysteine (Hcy) level, dietary intake of natural methylfolate, synthetic pteroylglutamic acid (PteGlu), vitamin C and C1561T-GCPII genotype. In controls and cases, 7.3 and 18.4 percent of subjects respectively, were found to have the CT genotype, increasing the risk for adenomatous polyp occurrence 2.86 times (95% CI:1.37-8.0, p=0.035). Total dietary folate, methylfolate and PteGlu intake and the level of erythrocyte folate and plasma Hcy did not predict the occurrence of an adenomatous polyp. However, dietary natural vitamin C intake was associated with adenomatous polyp risk within C1561T-GCPII CT genotype subjects (p=0.037). The findings suggest that C1561T-GCPII variation may be associated with risk for adenomatous polyp, and vitamin C may modify risk by interacting with the variant gene, its expression product and/or folate substrates.