Přejít k obsahu
Merck
Integrin LFA-1 regulates cell adhesion via transient clutch formation.
Biochemical and biophysical research communications (2015-07-06)
Munenori Ishibashi, Yukihiro Miyanaga, Satomi Matsuoka, Jun Kozuka, Yuichi Togashi, Tatsuo Kinashi, Masahiro Ueda
ANOTACE
Integrin LFA-1 regulates immune cell adhesion and trafficking by binding to ICAM-1 upon chemokine stimulation. Integrin-mediated clutch formation between extracellular ICAM-1 and the intracellular actin cytoskeleton is important for cell adhesion. We applied single-molecule tracking analysis to LFA-1 and ICAM-1 in living cells to examine the ligand-binding kinetics and mobility of the molecular clutch under chemokine-induced physiological adhesion and Mn(2+)-induced tight adhesion. Our results show a transient LFA-1-mediated clutch formation that lasts a few seconds and leads to a transient lower-mobility is sufficient to promote cell adhesion. Stable clutch formation was observed for Mn(2+)-induced high affinity LFA-1, but was not required for physiological adhesion. We propose that fast cycling of the clutch formation by intermediate-affinity integrin enables dynamic cell adhesion and migration.