Přejít k obsahu
Merck
  • Gene expression profiling in ethnic Malays with type 2 diabetes mellitus, with and without diabetic nephropathy.
Gene expression profiling in ethnic Malays with type 2 diabetes mellitus, with and without diabetic nephropathy.
Journal of nephrology (2011-03-02)
Faradianna E Lokman, Norhashimah A Seman, Aziz Al-Safi Ismail, Nor Azwany Yaacob, Norlaila Mustafa, Amir S M Khir, Zanariah Hussein, Wan Nazaimoon Wan Mohamud
ANOTACE
Diabetic nephropathy (DN) is the most common cause of end-stage renal disease (ESRD) among type 2 diabetes mellitus patients (DM) in Malaysia. This study used microarray analysis to determine the gene expression profiling in ethnic Malay patients with type 2 DM. A total of 312 patients were recruited; 25 were on dialysis due to ESRD, 128 were classified as normoalbuminuric, 93 as microalbuminuric and 66 as macroalbuminuric, based on urine albumin to creatinine ratio of <3.5, between 3.5 and 35 and =35 mg/mmol, respectively. Microalbuminuria was associated with up- and down-regulation of 2,694 and 2,538 genes, respectively, while macroalbuminuria was associated with up-regulation of 2,520 genes and down-regulation of 2,920 genes. There was significant up-regulation of 1,135 genes and down-regulation of 908 genes in the ESRD samples. Thirty-seven significantly up-regulated genes and 40 down-regulated genes were commonly expressed in all 3 groups of patients with worsening of renal functions. Up-regulated genes included major histocompatibility complex (HLA-C), complement component 3a receptor 1 (C3AR1), solute carrier family 16, member 3 (SLC16A3) and solute carrier family 9 (sodium/hydrogen exchanger) (SLC9A8). Consistently down-regulated genes included were bone morphogenetic phosphatase kinase (BMP2K), solute carrier family 12, member 1 (SLC12A1), solute carrier family 7 (SLC7A2), paternally expressed 10 (PEG10) and protein phosphatase 1 regulatory (inhibitor unit) (PPP1R1C). This study has identified several genes of interest, such as HLA-C, SLC16A3, SLC9A8, SLC12A1 and SLC7A2, that require verification of their roles as susceptibility genes for diabetic nephropathy in ethnic Malays with type 2 DM.