Přejít k obsahu
Merck
  • Attenuation of microRNA-1 derepresses the cytoskeleton regulatory protein twinfilin-1 to provoke cardiac hypertrophy.
Attenuation of microRNA-1 derepresses the cytoskeleton regulatory protein twinfilin-1 to provoke cardiac hypertrophy.
Journal of cell science (2010-06-24)
Qing Li, Xiao-Wei Song, Jun Zou, Guo-Kun Wang, Elena Kremneva, Xiang-Qi Li, Ni Zhu, Tao Sun, Pekka Lappalainen, Wen-Jun Yuan, Yong-Wen Qin, Qing Jing
ANOTACE
MicroRNAs are involved in several aspects of cardiac hypertrophy, including cardiac growth, conduction, and fibrosis. However, their effects on the regulation of the cardiomyocyte cytoskeleton in this pathological process are not known. Here, with microRNA microarray and small RNA library sequencing, we show that microRNA-1 (miR-1) is the most abundant microRNA in the human heart. By applying bioinformatic target prediction, a cytoskeleton regulatory protein twinfilin-1 was identified as a potential target of miR-1. Overexpression of miR-1 not only reduced the luciferase activity of the reporter containing the 3' untranslated region of twinfilin-1 mRNA, but also suppressed the endogenous protein expression of twinfilin-1, indicating that twinfilin-1 is a direct target of miR-1. miR-1 was substantially downregulated in the rat hypertrophic left ventricle and phenylephrine-induced hypertrophic cardiomyocytes, and accordingly, the protein level of twinfilin-1 was increased. Furthermore, overexpression of miR-1 in hypertrophic cardiomyocytes reduced the cell size and attenuated the expression of hypertrophic markers, whereas silencing of miR-1 in cardiomyocytes resulted in the hypertrophic phenotype. In accordance, twinfilin-1 overexpression promoted cardiomyocyte hypertrophy. Taken together, our results demonstrate that the cytoskeleton regulatory protein twinfilin-1 is a novel target of miR-1, and that reduction of miR-1 by hypertrophic stimuli induces the upregulation of twinfilin-1, which in turn evokes hypertrophy through the regulation of cardiac cytoskeleton.