Přejít k obsahu
Merck
  • Antiviral action of synthetic stigmasterol derivatives on herpes simplex virus replication in nervous cells in vitro.
Antiviral action of synthetic stigmasterol derivatives on herpes simplex virus replication in nervous cells in vitro.
BioMed research international (2014-08-26)
Erina Petrera, Analía G Níttolo, Laura E Alché
ANOTACE
Polyfunctionalized stigmasterol derivatives, (22S,23S)-22,23-dihydroxystigmast-4-en-3-one (compound 1) and (22S,23S)-3β-bromo-5α,22,23-trihydroxystigmastan-6-one (compound 2), inhibit herpes simplex virus type 1 (HSV-1) replication and spreading in human epithelial cells derived from ocular tissues. Both compounds reduce the incidence and severity of lesions in a murine model of herpetic stromal keratitis when administered in different treatment modalities. Since encephalitis caused by HSV-1 is another immunopathology of viral origin, we evaluate here the antiviral effect of both compounds on HSV-1 infected nervous cell lines as well as their anti-inflammatory action. We found that both stigmasterol derivatives presented low cytotoxicity in the three nervous cell lines assayed. Regarding the antiviral activity, in all cases both compounds prevented HSV-1 multiplication when added after infection, as well as virus propagation. Additionally, both compounds were able to hinder interleukin-6 and Interferon-gamma secretion induced by HSV-1 infection in Neuro-2a cells. We conclude that compounds 1 and 2 have exerted a dual antiviral and anti-inflammatory effect in HSV-1 infected nervous cell lines, which makes them interesting molecules to be further studied.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Cotton Rat Ifng / Interferon Gamma ELISA Kit