Přejít k obsahu
Merck
  • The up-regulation of miR-199b-5p in erythroid differentiation is associated with GATA-1 and NF-E2.
The up-regulation of miR-199b-5p in erythroid differentiation is associated with GATA-1 and NF-E2.
Molecules and cells (2014-03-13)
Yuxia Li, Hua Bai, Zhongzu Zhang, Weihua Li, Lei Dong, Xueju Wei, Yanni Ma, Junwu Zhang, Jia Yu, Guotao Sun, Fang Wang
ANOTACE
MicroRNAs (miRNAs) represent a class of small non-coding regulatory RNAs that play important roles in normal hematopoiesis, including erythropoiesis. Although studies have identified several miRNAs that regulate erythroid commitment and differentiation, we do not understand the mechanism by which the crucial erythroid transcription factors, GATA-1and NF-E2 directly regulate and control differentiation via miRNA pathways. In this study, we identified miR-199b-5p as a key regulator of human erythropoiesis, and its expression was up-regulated during the erythroid differentiation of K562 cells. Furthermore, the increase of miR-199b-5p in erythroid cells occurred in a GATA-1- and NF-E2-dependent manner during erythrocyte maturation. Both GATA-1 and NF-E2 bound upstream of the miR-199b gene locus and activated its transcription. Forced expression of miRNA-199b-5p in K562 cells affected erythroid cell proliferation and maturation. Moreover, we identified c-Kit as a direct target of miR-199b-5p in erythroid cells. Taken together, our results establish a functional link among the erythroid transcription factors GATA-1/NF-E2, miR-199b-5p and c-Kit, and provide new insights into the coupling of transcription and post-transcription regulation in erythroid differentiation.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Anti-NF-E2-related factor 2 Antibody, from rabbit, purified by affinity chromatography