Přejít k obsahu
Merck
cNMP-AMs mimic and dissect bacterial nucleotidyl cyclase toxin effects.
Biochemical and biophysical research communications (2014-08-12)
Ulrike Beckert, Manuel Grundmann, Sabine Wolter, Frank Schwede, Holger Rehmann, Volkhard Kaever, Evi Kostenis, Roland Seifert
ANOTACE
In addition to the well-known second messengers cAMP and cGMP, mammalian cells contain the cyclic pyrimidine nucleotides cCMP and cUMP. The Pseudomonas aeruginosa toxin ExoY massively increases cGMP and cUMP in cells, whereas the Bordetella pertussis toxin CyaA increases cAMP and, to a lesser extent, cCMP. To mimic and dissect toxin effects, we synthesized cNMP-acetoxymethylesters as prodrugs. cNMP-AMs rapidly and effectively released the corresponding cNMP in cells. The combination of cGMP-AM plus cUMP-AM mimicked cytotoxicity of ExoY. cUMP-AM and cGMP-AM differentially activated gene expression. Certain cCMP and cUMP effects were independent of the known cNMP effectors protein kinases A and G and guanine nucleotide exchange factor Epac. In conclusion, cNMP-AMs are useful tools to mimic and dissect bacterial nucleotidyl cyclase toxin effects.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Rp-8-Bromo-β-phenyl-1,N2-ethenoguanosine 3′,5′-cyclic monophosphorothioate sodium salt, ≥98% (HPLC), powder