Přejít k obsahu
Merck
  • Protein tyrosine phosphatase ζ enhances proliferation by increasing β-catenin nuclear expression in VHL-inactive human renal cell carcinoma cells.
Protein tyrosine phosphatase ζ enhances proliferation by increasing β-catenin nuclear expression in VHL-inactive human renal cell carcinoma cells.
World journal of urology (2013-04-17)
Donghao Shang, Xiuhong Xu, Daye Wang, Yong Li, Yuting Liu
ANOTACE
We investigated the role of protein tyrosine phosphatase ζ (Ptprz1) in human renal cell carcinoma (RCC) cells' proliferation and associations between Ptprz1 expression and von Hippel-Lindau (VHL) activation. A normal human renal cell line and four human RCC cell lines were used in this study. VHL or Ptprz1 expression in RCC cells was increased by transfection with a VHL or Ptprz1 vector. VHL or Ptprz1 expression was decreased in these cells by siRNA using Lipofectamine 2000. Cells' proliferative activity was assessed by WST-1 assay. Our results suggested that Ptprz1 was a target of VHL, and a loss of VHL activation increased Ptprz1 expression in RCC cells. Ptprz1 enhanced β-catenin protein expressions in the nuclear fractions of RCC cells and participated in regulating proliferation by activating β-catenin and its downstream genes. In addition, a loss of VHL activity may enhance the proliferative activity of RCC cells by increasing Ptprz1 expression. Ptprz1-enhanced RCC cells' proliferation depends on VHL inactivation, and the Ptprz1/β-catenin pathway may be a potential target for treating RCC with inactive VHL.