Přejít k obsahu
Merck
  • Quantitative real-time RT-PCR of ITGA7, SVEP1, TNS1, LPHN3, SEMA3G, KLB and MMP13 mRNA expression in breast cancer.
Quantitative real-time RT-PCR of ITGA7, SVEP1, TNS1, LPHN3, SEMA3G, KLB and MMP13 mRNA expression in breast cancer.
Asian Pacific journal of cancer prevention : APJCP (2013-01-16)
Manas Kotepui, Charin Thawornkuno, Porntip Chavalitshewinkoon-Petmitr, Phaibul Punyarit, Songsak Petmitr
ANOTACE
Breast cancer is the leading cause of cancer deaths among women worldwide, including Thailand. In the present study, the differential mRNA expression of SVEP1, LPHN3, KLB, ITGA7, SEMA3G, TNS1 and MMP13 genes was examined in breast cancer using quantitative real-time reverse transcription polymerase chain reaction (QRT-PCR). Among these genes, increased LPHN3 and MMP13 mRNA expression levels correlated with axillary-node metastasis (P=0.02). Multiple logistic regression analysis revealed that LPHN3 and MMP13 mRNA expression is significantly associated with axillary node status in breast cancer (P=0.04).