Přejít k obsahu
Merck
  • GOA, a novel gene encoding a ring finger B-box coiled-coil protein, is overexpressed in astrocytoma.
GOA, a novel gene encoding a ring finger B-box coiled-coil protein, is overexpressed in astrocytoma.
Biochemical and biophysical research communications (2001-08-21)
D A Vandeputte, C B Meije, M van Dartel, S Leenstra, H IJlst-Keizers, P K Das, D Troost, D A Bosch, F Baas, T J Hulsebos
ANOTACE
Serial analysis of gene expression (SAGE) was used to identify a gene named GOA (gene overexpressed in astrocytoma), which codes for a novel Ring finger B-box coiled-coil (RBCC) protein. Northern blot hybridization showed overexpression of GOA in 9 of 10 astrocytomas. Except for kidney, in which high expression was found, expression levels in normal tissues were low and comparable to normal brain. Immunohistochemistry demonstrated presence of GOA, with prominent nuclear staining, in astrocytoma tumor cells and astrocytes of fetal brain, but virtual absence in mature astrocytes. Overexpression was not due to amplification, since amplification of GOA was only found in one of 65 astrocytomas. GOA was localized to 17q24-25, a region that is frequently gained or amplified in a number of other tumor types. GOA contains two LXXLL motifs, which are thought to be important for nuclear receptor binding. Our data suggest an important role of GOA in the process of dedifferentiation that is associated with astrocytoma tumorigenesis and possibly with that of other tumor types as well.