Přejít k obsahu
Merck
  • Altered laminin 5 expression due to mutations in the gene encoding the beta 3 chain (LAMB3) in generalized atrophic benign epidermolysis bullosa.
Altered laminin 5 expression due to mutations in the gene encoding the beta 3 chain (LAMB3) in generalized atrophic benign epidermolysis bullosa.
The Journal of investigative dermatology (1995-04-01)
J A McGrath, L Pulkkinen, A M Christiano, I M Leigh, R A Eady, J Uitto
ANOTACE
The anchoring filament component laminin 5 (kalinin/nicein) is a candidate protein for mutations in some hereditary blistering skin disorders. In this study, laminin 5 expression was assessed in a family with generalized atrophic benign epidermolysis bullosa, a non-lethal variant of the junctional form of epidermolysis bullosa. Immunofluorescence microscopy of the skin basement-membrane zone with a monoclonal antibody (GB3) revealed reduced anti-laminin 5 staining compared to normal controls. The labeling, when examined by immunoelectron microscopy, was present within the lower lamina lucida, immediately below the plane of blister formation. Numerous hemidesmosomes and well-formed anchoring filaments were seen on transmission electron microscopy. Polymerase chain reaction amplification of genomic DNA encoding the beta 3 subunit (LAMB3) of laminin 5, heteroduplex analysis of the polymerase chain reaction products, and nucleotide sequencing of the heteroduplexes revealed two putative mutations within the LAMB3 gene; these consisted of a premature termination codon in exon 3 and a missense mutation in exon 7. Exons 3 and 7 encode part of domain VI of the laminin 5 beta 3 chain short arm. This globular domain of the protein has been postulated to have an important function in the interaction of laminin 5 with other structural components of the basement membrane zone, such as laminin 6 (K-laminin). Thus the mutations delineated in this family may have a critical pathogenetic significance in reducing adhesion between the epidermis and the dermis.