Přejít k obsahu
Merck
  • Three novel sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) 3 isoforms. Expression, regulation, and function of the membranes of the SERCA3 family.
Three novel sarco/endoplasmic reticulum Ca2+-ATPase (SERCA) 3 isoforms. Expression, regulation, and function of the membranes of the SERCA3 family.
The Journal of biological chemistry (2002-04-17)
Virginie Martin, Raymonde Bredoux, Elisabeth Corvazier, Roosje Van Gorp, Tunde Kovacs, Pascal Gelebart, Jocelyne Enouf
ANOTACE
Sarco/endoplasmic reticulum Ca2+-ATPases (SERCAs) pump Ca2+ into the endoplasmic reticulum. Recently, three human SERCA3 (h3a-c) proteins and a previously unknown rat SERCA3 (r3b/c) mRNA have been described. Here, we (i) document two novel human SERCA3 splice variants h3d and h3e, (ii) provide data for the expression and mechanisms regulating the expression of all known SERCA3 variants (r3a, r3b/c, and h3a-e), and (iii) show functional characteristics of the SERCA3 isoforms. h3d and h3e are issued from the insertion of an additional penultimate exon 22 resulting in different carboxyl termini for these variants. Distinct distribution patterns of the SERCA3 gene products were observed in a series of cell lines of hematopoietic, epithelial, embryonic origin, and several cancerous types, as well as in panels of rat and human tissues. Hypertension and protein kinase C, calcineurin, or retinoic acid receptor signaling pathways were found to differently control rat and human splice variant expression, respectively. Stable overexpression of each variant was performed in human embryonic kidney 293 cells, and the SERCA3 isoforms were fully characterized. All SERCA3 isoforms were found to pump Ca2+ with similar affinities. However, they modulated the cytosolic Ca2+ concentration ([Ca2+]c) and the endoplasmic reticulum Ca2+ content ([Ca2+]er) in different manners. A newly generated polyclonal antibody and a pan-SERCA3 antibody proved the endogenous expression of the three novel SERCA3 proteins, h3d, h3e, and r3b/c. All these data suggest that the SERCA3 gene products have a more widespread role in cellular Ca2+ signaling than previously appreciated.