- Nonsteroidal progesterone receptor ligands. 2. High-affinity ligands with selectivity for bone cell progesterone receptors.
Nonsteroidal progesterone receptor ligands. 2. High-affinity ligands with selectivity for bone cell progesterone receptors.
Journal of medicinal chemistry (1995-12-08)
D W Combs, K Reese, L A Cornelius, J W Gunnet, E V Cryan, K S Granger, J J Jordan, K T Demarest
PMID8523400
ANOTACE
A novel series of nonsteroidal heterocycles was discovered which display cell-type selective, high-affinity (nanomolar) binding to the progesterone receptors from TE85 osteosarcoma cells but > 1 microM binding affinity to the progesterone receptors from T47D and ZR75 human breast carcinoma cells. Structure-activity relationships were developed for a set of these compounds, and a representative analog 1-(3,4-dichlorobenzoyl)-3-phenyl-1,4,5,6-tetrahydropyridazine++ + (1i, RWJ 25333) was chosen for further evaluation. RWJ 25333 stimulated the in vitro proliferation of human osteoblast-like cells but not human breast cells.
MATERIÁLY