Přejít k obsahu
Merck
  • Anionic heptadecanuclear silver(I) cluster constructed from in situ generated 2-mercaptobenzoic acid and a sulfide anion.
Anionic heptadecanuclear silver(I) cluster constructed from in situ generated 2-mercaptobenzoic acid and a sulfide anion.
Inorganic chemistry (2011-11-11)
Di Sun, Fu-Jing Liu, Rong-Bin Huang, Lan-Sun Zheng
ANOTACE
A novel anionic heptadecanuclear silver(I) cluster, (NH(4))(17)[(μ(6)-S)@Ag(17)(mba)(16)]·22H(2)O (1; H(2)mba = 2-mercaptobenzoic acid), was obtained by the reaction of equivalent molar silver oxide and 2,2'-dithiodibenzoic acid (H(2)dtba) under ultrasonic conditions at 50 °C. Complex 1 is a discrete cluster comprised of unexpected mba ligands on the shell and a μ(6)-S(2-) ion in the core, suggesting the occurrence of in situ S-S and S-C(sp(2)) bond cleavages of the H(2)dtba ligand. This novel cluster displays moderate orange-red emission in the solid state at room temperature.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
2,2′-Dithiodibenzoic acid, ≥95.0% (NT)