Přejít k obsahu
Merck
  • Association of serum arginase I with L-arginine, 3-nitrotyrosine, and exhaled nitric oxide in healthy Japanese workers.
Association of serum arginase I with L-arginine, 3-nitrotyrosine, and exhaled nitric oxide in healthy Japanese workers.
Free radical research (2013-09-26)
K Ogino, D H Wang, M Kubo, Y Obase, H Setiawan, F Yan, H Takahashi, R Zhang, Y Tsukiyama, J Yoshida, Y Zou
ANOTACE
The associations of serum arginase I with serum L-arginine, serum 3-nitrotyrosine, and fractional exhaled nitric oxide (FENO) were evaluated cross-sectionally in healthy Japanese workers. The serum median (minimum-maximum) levels of arginase I, 3-nitrotyrosine, and FENO in healthy people (n = 130) were 14.6 (0.94-108.1) ng/mL, 81.0 (0.27-298.6) pmol/mg protein, and 14.0 (5.0-110.0) parts per billion, respectively. Significant correlations of arginase I with FENO, L-arginine, 3-nitrotyrosine, and percent predicted forced expiratory volume in 1 s (FEV1 (% predicted)) were observed, and correlations of FENO with immunoglobulin E (IgE), NOx, arginase I, and sex and allergy were also observed. By multiple regression analysis, arginase I showed positive associations with FENO and 3-nitrotyrosine, and a negative association with L-arginine; and FENO showed positive associations with IgE and NO2(-) + NO3(-) (NOx), and a negative association with L-arginine, as well as an association with sex. Moreover, logistic regression analysis showed linear inverse associations of arginase I and 3-nitrotyrosine with L-arginine, and showed linear positive associations of FENO with IgE and NOx. It was concluded that serum arginase I might regulate serum L-arginine and 3-nitrotyrosine via L-arginine, and that IgE or NOx might regulate FENO in a healthy Japanese population.