Přejít k obsahu
Merck
The structure of dimeric apolipoprotein A-IV and its mechanism of self-association.
Structure (London, England : 1993) (2012-05-15)
Xiaodi Deng, Jamie Morris, James Dressmen, Matthew R Tubb, Patrick Tso, W Gray Jerome, W Sean Davidson, Thomas B Thompson
ANOTACE
Apolipoproteins are key structural elements of lipoproteins and critical mediators of lipid metabolism. Their detergent-like properties allow them to emulsify lipid or exist in a soluble lipid-free form in various states of self-association. Unfortunately, these traits have hampered high-resolution structural studies needed to understand the biogenesis of cardioprotective high-density lipoproteins (HDLs). We derived a crystal structure of the core domain of human apolipoprotein (apo)A-IV, an HDL component and important mediator of lipid absorption. The structure at 2.4 Å depicts two linearly connected 4-helix bundles participating in a helix swapping arrangement that offers a clear explanation for how the protein self-associates as well as clues to the structure of its monomeric form. This also provides a logical basis for antiparallel arrangements recently described for lipid-containing particles. Furthermore, we propose a "swinging door" model for apoA-IV lipid association.