Přejít k obsahu
Merck
  • Functional interaction between SHPTP1 and the Lyn tyrosine kinase in the apoptotic response to DNA damage.
Functional interaction between SHPTP1 and the Lyn tyrosine kinase in the apoptotic response to DNA damage.
The Journal of biological chemistry (1999-11-27)
K Yoshida, S Kharbanda, D Kufe
ANOTACE
The Lyn protein-tyrosine kinase is activated in the cellular response to DNA-damaging agents. Here we demonstrate that Lyn associates constitutively with the SHPTP1 protein-tyrosine phosphatase. The SH3 domain of Lyn interacts directly with SHPTP1. The results show that Lyn phosphorylates SHPTP1 at the C-terminal Tyr-564 site. Lyn-mediated phosphorylation of SHPTP1 stimulates SHPTP1 tyrosine phosphatase activity. We also demonstrate that treatment of cells with 1-beta-D-arabinofuranosylcytosine and other genotoxic agents induces Lyn-dependent phosphorylation and activation of SHPTP1. The significance of the Lyn-SHPTP1 interaction is supported by the demonstration that activation of Lyn contributes in part to the apoptotic response to ara-C treatment and that SHPTP1 attenuates this response. These findings support a functional interaction between Lyn and SHPTP1 in the response to DNA damage.