Přejít k obsahu
Merck
  • Stereoselective synthesis of highly functionalized nitrocyclopropanes via organocatalyic conjugate addition to nitroalkenes.
Stereoselective synthesis of highly functionalized nitrocyclopropanes via organocatalyic conjugate addition to nitroalkenes.
The Journal of organic chemistry (2006-09-09)
Séamus H McCooey, Thomas McCabe, Stephen J Connon
ANOTACE
A convenient and novel one-pot organocatalytic methodology for the stereoselective synthesis of highly functionalized nitrocyclopropanes is reported. The addition of dimethyl chloromalonate to a variety of nitroolefins catalyzed by tertiary amines leads to a Michael adduct which cyclizes to form the cyclopropane in the presence of DBU under carefully controlled reaction conditions with outstanding diastereoselectivity.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Dimethyl chloromalonate, 94%