Přejít k obsahu
Merck
  • PCOS is associated with increased CD11c expression and crown-like structures in adipose tissue and increased central abdominal fat depots independent of obesity.
PCOS is associated with increased CD11c expression and crown-like structures in adipose tissue and increased central abdominal fat depots independent of obesity.
The Journal of clinical endocrinology and metabolism (2012-11-03)
Zhi Hua Huang, Buvana Manickam, Victoria Ryvkin, Xiaohong Joe Zhou, Giamila Fantuzzi, Theodore Mazzone, Susan Sam
ANOTACE
Adipose tissue macrophage (ATM) infiltration is a major pathway for obesity-induced insulin resistance but has not been studied as a mechanism for insulin resistance in PCOS. We tested whether polycystic ovary syndrome (PCOS) is associated with increased ATM infiltration, especially of inflammatory subtype identified by the CD11c marker. We conducted a case-control study at an academic medical center in the United States. Fourteen PCOS and 14 control women of similar age and body mass index (BMI) underwent a gluteal fat biopsy. Markers of ATM, integrins, TNF-α, and adiponectin, were analyzed by quantitative RT-PCR using a standard curve method. Crown-like structures (CLS) were identified by immunohistochemistry. Abdominal magnetic resonance imaging and frequently sampled i.v. glucose tolerance test were performed to assess abdominal fat and insulin sensitivity (SI). Women with PCOS were compared with control women of similar age and BMI for ATM markers, CLS density, adipose tissue expression of inflammatory cytokines and adiponectin, SI, and abdominal fat depots. Women with PCOS had an increase in CD11c expression (P = 0.03), CLS density (P = 0.001), α5 expression (P = 0.009), borderline increase in TNF-α expression (P = 0.08), and a decrease in adiponectin expression (P = 0.02) in gluteal adipose tissue. Visceral (P = 0.009) and sc abdominal fat (P = 0.005) were increased in PCOS. SI was lower in PCOS (P = 0.008). PCOS is associated with an increase in CD11c expression and CLS density and a decrease in adiponectin expression in sc adipose tissue. Additionally, PCOS is associated with higher central abdominal fat depots independent of BMI. These alterations are present among mostly nonobese women and could represent mechanisms for insulin resistance.