Přejít k obsahu
Merck
EB1 reveals mobile microtubule nucleation sites in Arabidopsis.
Nature cell biology (2003-10-15)
Jordi Chan, Grant M Calder, John H Doonan, Clive W Lloyd
ANOTACE
In plants, it is unclear how dispersed cortical microtubules are nucleated, polarized and organized in the absence of centrosomes. In Arabidopsis thaliana cells, expression of a fusion between the microtubule-end-binding protein AtEB1a and green fluorescent protein (GFP) results in labelling of spindle poles, where minus ends gather. During interphase, AtEB1a-GFP labels the microtubule plus end as a comet, but also marks the minus end as a site from which microtubules can grow and shrink. These minus-end nucleation sites are mobile, explaining how the cortical array can redistribute during the cell cycle and supporting the idea of a flexible centrosome in plants.