- Gravure printing of graphene for large-area flexible electronics.
Gravure printing of graphene for large-area flexible electronics.
Advanced materials (Deerfield Beach, Fla.) (2014-05-02)
Ethan B Secor, Sooman Lim, Heng Zhang, C Daniel Frisbie, Lorraine F Francis, Mark C Hersam
PMID24782064
ANOTACE
Gravure printing of graphene is demonstrated for the rapid production of conductive patterns on flexible substrates. Development of suitable inks and printing parameters enables the fabrication of patterns with a resolution down to 30 μm. A mild annealing step yields conductive lines with high reliability and uniformity, providing an efficient method for the integration of graphene into large-area printed and flexible electronics.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Graphene ink, for gravure printing, with ethyl cellulose in terpineol, gravure printable