Přejít k obsahu
Merck
  • Propentofylline: a nucleoside transport inhibitor with neuroprotective effects in cerebral ischemia.
Propentofylline: a nucleoside transport inhibitor with neuroprotective effects in cerebral ischemia.
General pharmacology (1994-10-01)
F E Parkinson, K A Rudolphi, B B Fredholm
ANOTACE
1. Adenosine is an endogenous neuroprotective agent; stimulation of A1 receptors decreases excitatory amino acid neurotransmission and stimulation of A2 receptors inhibits platelet and neutrophil activation and promotes vasodilation. 2. Post-ischemic administration of propentofylline (HWA 285) reduces neuronal damage in gerbils and improves glucose metabolism in all regions of brain in acute stroke patients. 3. Propentofylline inhibits the transport of adenosine into cultured cells and increases extracellular adenosine concentrations in ischemic brain. Thus, enhanced stimulation of adenosine receptors may account for some of the neuroprotective effects of this compound. 4. Propentofylline inhibits free radical production by cultivated microglia cells, stimulates nerve growth factor production and inhibits cAMP-phosphodiesterase activity. These effects may also be important for neuroprotection.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Propentofylline, solid