Přejít k obsahu
Merck
Efficacy of isosorbide mononitrate in angina pectoris.
The American journal of cardiology (1992-11-27)
U Thadani, P J de Vane
ANOTACE
The rapid development of tolerance has limited the applicability of oral and transdermal nitrates in the long-term management of patients with chronic stable angina pectoris. Recent well-controlled trials have demonstrated that asymmetrical, or eccentric, dosing of oral isosorbide mononitrate, in which 20-mg doses are taken at 8 A.M. and 3 P.M., provides at least 12 hours of antianginal coverage. There is no evidence for the development of tolerance with this schedule, which allows for a 17-hour nitrate withdrawal period. Likewise, the asymmetrical 20-mg twice daily regimen has not been associated with the zero-hour effect that has been reported with higher oral doses of isosorbide mononitrate and with intermittent nitroglycerin patch therapy. This approach also avoids the development of a clinical rebound phenomenon, as measured by increased episodes of angina and nitroglycerin consumption, compared with the pretreatment period, during the nitrate-free interval at night and the early hours of the morning.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Isosorbide mononitrate, European Pharmacopoeia (EP) Reference Standard