Přejít k obsahu
Merck
  • Vibrational (FT-IR and FT-Raman) spectra, NBO, HOMO-LUMO, Molecular electrostatic potential surface and computational analysis of 4-(trifluoromethyl)benzylbromide.
Vibrational (FT-IR and FT-Raman) spectra, NBO, HOMO-LUMO, Molecular electrostatic potential surface and computational analysis of 4-(trifluoromethyl)benzylbromide.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2013-08-06)
M Karnan, V Balachandran, M Murugan, M K Murali, A Nataraj
ANOTACE
In this work, the vibrational characteristics of 4-(trifluoromethyl) benzylbromide (TFMBB) have been investigated and both the experimental and theoretical vibrational data indicate the presence of functional groups in the title molecule. The density functional theoretical (DFT) computations were performed at the B3LYP/6-311+G (d,p) levels to derive the optimized geometry, vibrational wavenumbers with IR and Raman intensities. Furthermore, the molecular orbital calculations such as natural bond orbitals (NBOs), HOMO-LUMO energy gap and Mapped molecular electrostatic potential (MEP) surfaces were also performed with the same level of DFT. The thermal flexibility of molecule in associated with vibrational temperature was also illustrated on the basis of correlation graphs. The detailed interpretation of the vibrational spectra has been carried out with the aid of potential energy distribution (PED) results obtained from MOLVIB program. The delocalization of electron density of various constituents of the molecule has been discussed with the aid of NBO analysis.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
4-(Trifluoromethyl)benzyl bromide, 98%