Přejít k obsahu
Merck
  • Analysis of anthocyanin pigments in Lonicera (Caerulea) extracts using chromatographic fractionation followed by microcolumn liquid chromatography-mass spectrometry.
Analysis of anthocyanin pigments in Lonicera (Caerulea) extracts using chromatographic fractionation followed by microcolumn liquid chromatography-mass spectrometry.
Journal of chromatography. A (2010-11-30)
Renáta Myjavcová, Petr Marhol, Vladimír Křen, Vilím Simánek, Jitka Ulrichová, Irena Palíková, Barbora Papoušková, Karel Lemr, Petr Bednář
ANOTACE
Anthocyanins from the fruit Lonicera caerulea L. var. kamtschatica (blueberry honeysuckle, Caprifoliaceae) were studied via (semi)preparative chromatographic fractionation followed by MS and μLC/MS analysis. The extraction procedure was optimized with respect to analytical purposes as well as its potential use for the preparation of nutraceuticals. The highest yield of anthocyanins was obtained using acidified methanol as the extraction medium. A comparable total anthocyanin content was obtained using a mixture of methanol and acetone. However, when Lonicera anthocyanins were in contact with acetone, a condensation reaction occurred to a large extent and related 5-methylpyranoanthocyanins were found. The effect of other extraction media, including ethanol as a "green" solvent, is also discussed. The potential of two fractionation procedures for extract purification differing in their chromatographic selectivity and scale was studied (i.e. using a Sephadex LH-20 gel column and a reversed phase). Fractions obtained by both procedures were used for a detailed analysis. MS and μLC/MS(2) methods were used for monitoring anthocyanin and 5-methylpyranoderivatives content as well as identifying less common and more complex dyes (dimer of cyanidin-3-hexoside, cyanidin-ethyl-catechin-hexosides, etc.). These more complex dyes are most likely formed during fruit treatment.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Supelco
Sepabeads® SP-207, bucket of 1000 g