Přejít k obsahu
Merck
  • Thyroid hormone receptor interacting protein 3 (trip3) is a novel coactivator of hepatocyte nuclear factor-4alpha.
Thyroid hormone receptor interacting protein 3 (trip3) is a novel coactivator of hepatocyte nuclear factor-4alpha.
Diabetes (2002-03-28)
Hiromi Iwahashi, Kazuya Yamagata, Issei Yoshiuchi, Jungo Terasaki, Qin Yang, Kenji Fukui, Arisa Ihara, Qian Zhu, Takeshi Asakura, Yang Cao, Akihisa Imagawa, Mitsuyoshi Namba, Toshiaki Hanafusa, Jun-ichiro Miyagawa, Yuji Matsuzawa
ANOTACE
Mutations of the hepatocyte nuclear factor-4alpha (HNF-4alpha) gene are associated with a subtype of maturity-onset diabetes of the young (MODY1) that is characterized by impaired insulin secretion in response to a glucose load. HNF-4alpha, which is a transcription factor expressed in pancreatic beta-cells, plays an important role in regulating the expression of genes involved in glucose metabolism. Thus, cofactors that interact with HNF-4alpha and modify its transcriptional activity might also play an important role in regulating the metabolic pathways in pancreatic beta-cells, and the genes of such cofactors are plausible candidate genes for MODY. In the present study, we showed, using a yeast two-hybrid screening assay, that thyroid hormone receptor interacting protein 3 (Trip3) interacted with HNF-4alpha, and their interaction was confirmed by the glutathione S-transferase pull-down assay. Human Trip3 cDNA contained an open reading frame for a protein of 155 amino acids, and the gene was expressed in both pancreatic islets and MIN6 cells. Cotransfection experiments indicated that Trip3 could enhance (two- to threefold) the transcription activity of HNF-4alpha in COS-7 cells and MIN6 cells. These results suggest that Trip3 is a coactivator of HNF-4alpha. Mutation screening revealed that variation of the Trip3 gene is not a common cause of MODY/early-onset type 2 diabetes in Japanese individuals. Trip3 may play an important role in glucose metabolism by regulating the transcription activity of HNF-4alpha.