Přejít k obsahu
Merck
  • Genetic variants in FGFR2 and MAP3K1 are associated with the risk of familial and early-onset breast cancer in a South-American population.
Genetic variants in FGFR2 and MAP3K1 are associated with the risk of familial and early-onset breast cancer in a South-American population.
Breast cancer research and treatment (2012-12-12)
Lilian Jara, Patricio Gonzalez-Hormazabal, Kerube Cerceño, Gabriella A Di Capua, Jose M Reyes, Rafael Blanco, Teresa Bravo, Octavio Peralta, Fernando Gomez, Enrique Waugh, Sonia Margarit, Gladys Ibañez, Carmen Romero, Janara Pakomio, Gigia Roizen
ANOTACE
Genome-Wide Association Studies have identified several loci associated with breast cancer (BC) in populations of different ethnic origins. One of the strongest associations was found in the FGFR2 gene, and MAP3K1 has been proposed as a low-penetrance BC risk factor. In this study, we evaluated the associations among FGFR2 SNPs rs2981582, rs2420946, and rs1219648; and MAP3K1 rs889312, with BC risk in 351 BRCA1/2-negative Chilean BC cases and 802 controls. All the SNPs studied were significantly associated with increased BC risk in familial BC and in non-familial early-onset BC, in a dose-dependent manner. Subjects with 3 risk alleles were at a significantly increased risk of BC compared with subjects with 0-2 risk alleles, in both familial BC and early-onset non-familial BC (OR = 1.47, 95 % CI 1.04-2.07, P = 0.026 and OR = 2.04 95 % CI 1.32-3.24, P < 0.001, respectively). In the haplotype analysis, the FGFR2 rs2981582 T / rs2420946 T / rs1219648 G haplotype (ht2) was associated with a significantly increased BC risk compared with the rs2981582 C / rs2420946 C / rs1219648 A haplotype in familial BC and in non-familial early-onset BC (OR = 1.32, 95 % CI 1.06-1.65, P = 0.012; OR = 1.46, 95 % CI 1.11-1.91, P = 0.004, respectively). When the FGFR2 ht2 and MAP3K1 rs889312 were evaluated as risk alleles, the risk of BC increased in a dose-dependent manner as the number of risk alleles increased (P trend <0.0001), indicating an additive effect. Nevertheless, there is no evidence of an interaction between FGFR2 ht2 and the MAP3K1 rs889312 C allele. These findings suggest that genetic variants in the FGFR2 and MAP3K1 genes may contribute to genetic susceptibility to BC.