Přejít k obsahu
Merck
  • Small bowel preparations for capsule endoscopy with mannitol and simethicone: a prospective, randomized, clinical trial.
Small bowel preparations for capsule endoscopy with mannitol and simethicone: a prospective, randomized, clinical trial.
Journal of clinical gastroenterology (2010-09-28)
Hong-bin Chen, Yue Huang, Su-yu Chen, Hui-wen Song, Xiao-lin Li, Dong-lin Dai, Jia-tia Xie, Song He, Yuan-yuan Zhao, Chun Huang, Sheng-jun Zhang, Lin-na Yang
ANOTACE
There is no consensus concerning small bowel preparation before capsule endoscopy (CE). This study evaluated the effects of 4 regimens on small bowel cleansing and diagnostic yield. Patients were randomly divided into 4 groups. Group A consumed a clear liquid diet after lunch on the day before CE, followed by overnight fasting. Group B took 250 mL 20% mannitol and 1 L 0.9% saline orally at 05:00 hours on the day of the procedure. In group C, the same regimen was taken at 20:00 hours on the day before and at 05:00 hours on the day of CE. In group D, in addition to the group C regimen, 20 mL oral simethicone was taken 30 minutes before CE. Two hundred patients were prospectively enrolled, and 7 were excluded from the final analysis because of incomplete small bowel transit. No significant difference was noted among the 4 groups for small bowel transit time. Bowel preparation in group D was significantly better than for the other regimens for overall cleansing of the proximal small bowel, and showed improved overall cleansing of the distal small bowel when compared with 10-hours overnight fasting. Pathological lesions of the proximal and distal small bowel were, respectively, achieved in 82 and 74 patients, mostly distributed in group D. Small bowel preparation that involves split-dose oral mannitol plus single-dose simethicone for CE can improve mucosal visualization and subsequent diagnostic yield when compared with 10-hours overnight fasting.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
USP
Simethicone, United States Pharmacopeia (USP) Reference Standard