Přejít k obsahu
Merck
  • Proteomic analysis of testis biopsies in men treated with injectable testosterone undecanoate alone or in combination with oral levonorgestrel as potential male contraceptive.
Proteomic analysis of testis biopsies in men treated with injectable testosterone undecanoate alone or in combination with oral levonorgestrel as potential male contraceptive.
Journal of proteome research (2008-08-16)
Yugui Cui, Hui Zhu, Yefei Zhu, Xuejiang Guo, Ran Huo, Xinghai Wang, Jiansun Tong, Lixin Qian, Zuomin Zhou, Yue Jia, Yan-He Lue, Amiya Sinha Hikim, Christina Wang, Ronald S Swerdloff, Jiahao Sha
ANOTACE
Treatment with injectable testosterone undecanoate (TU) alone or in combination with oral levonorgestrel (LNG) resulted in marked decreases in sperm concentrations. In this study, we used proteomic analyses to examine the cellular/molecular events occurring in the human testis after TU or TU + LNG treatment. We conducted a global proteomic analysis of the human testicular biopsies before and at 2 weeks after TU alone or TU + LNG treatment. Proteins showing significant changes in expression were identified and analyzed. As a result, 17 and 46 protein spots were found with significant differential expression after the treatment with TU alone and TU + LNG, respectively. TU treatment changed the expression of heterogeneous nuclear ribonucleoprotein K (hnRNP K), proteasome inhibitor PI31 subunit (PSMF1), and superoxide dismutase [Mn] mitochondrial precursor (SOD2). These proteins inhibit "assembly", induce cell death, and promote compensatory "cell survival" in the testis. After TU + LNG treatment, "proliferation/cell survival" and "apoptosis/death" were the predominant responses in the testis. TU + LNG treatment inhibited the expression of Prolyl 4-hydroxylase beta subunit (P4HB) and Annexin A2 (Annexin II). These proteins are involved in apoptosis and cell proliferation, respectively. TU + LNG treatment also enhanced the expression of SOD2 and Parvalbumin alpha (Pvalb). These two proteins may protect testicular cells against apoptosis/death and promote cell survival. In conclusion, TU and TU + LNG treatments suppress spermatogenesis through different pathways by changing the expression of different proteins. hnRNP K, PSMF1, SOD2, P4HB, Annexin II, and Pvalb, are key proteins that may be early molecular targets responsible for spermatogenesis suppression induced by hormone treatment.