Přejít k obsahu
Merck
Fragrance material review on 2-(3-methylphenyl) ethanol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2011-11-01)
J Scognamiglio, L Jones, C S Letizia, A M Api
ANOTACE
A toxicologic and dermatologic review of 2-(3-methylphenyl) ethanol when used as a fragrance ingredient is presented. 2-(3-Methylphenyl) ethanol is a member of the fragrance structural group Aryl Alkyl Alcohols and is a primary alcohol. The AAAs are a structurally diverse class of fragrance ingredients that includes primary, secondary, and tertiary alkyl alcohols covalently bonded to an aryl (Ar) group, which may be either a substituted or unsubstituted benzene ring. The common structural element for the AAA fragrance ingredients is an alcohol group -C-(R1)(R2)OH and generically the AAA fragrances can be represented as an Ar-C-(R1)(R2)OH or Ar-Alkyl-C-(R1)(R2)OH group. This review contains a detailed summary of all available toxicology and dermatology papers that are related to this individual fragrance ingredient and is not intended as a stand-alone document. A safety assessment of the entire Aryl Alkyl Alcohols will be published simultaneously with this document; please refer to Belsito et al. (2012) for an overall assessment of the safe use of this material and all other branched chain saturated alcohols in fragrances.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
3-Methylphenethyl alcohol, 98%