Přejít k obsahu
Merck
Combinatorial synthesis and sensorial properties of polyfunctional thiols.
Journal of agricultural and food chemistry (2001-11-21)
C Vermeulen, J Pellaud, L Gijs, S Collin
ANOTACE
Over the past few years, polyfunctional thiols present as trace components have been found to play a major role in many food flavors, due to their exceptionally low odor thresholds. Unfortunately, their presence in minute concentration (in ng/kg to a few microg/kg) and their high reactivity make it very difficult to extract and identify them. Furthermore, most of them are not yet commercially available. The aim of this work was to characterize the chromatographic and sensorial properties of 10 synthetic mercaptoketones and mercaptoalcohols. Combinatorial chemistry proved to be a very useful way to synthesize them rapidly. Sulfur-selective sulfur chemiluminescence detection chromatograms coupled with mass spectroscopy enabled the target compounds to be identified. Flavor profiles and best estimate gas chromatography lowest amount detected by sniffing (BE-GC-LOADS) values were further determined by GC-olfactometry. As expected, new, exceptionally odorant molecules (BE-GC-LOADS < 0.1 ng) were revealed by this unusual approach.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
5-Methyl-3-hexen-2-one, technical grade, 75%