Přejít k obsahu
Merck
  • Tetraaquabis[5-(4-pyridyl N-oxide)tetrazolato-kappaN(2)]cadmium: the intermediate in the synthesis of a tetrazole from a nitrile and an azide.
Tetraaquabis[5-(4-pyridyl N-oxide)tetrazolato-kappaN(2)]cadmium: the intermediate in the synthesis of a tetrazole from a nitrile and an azide.
Acta crystallographica. Section C, Crystal structure communications (2004-04-09)
Zhao Xiang Yu, Xiao Ping Wang, Yaoyu Feng
ANOTACE
The intermediate [Cd(4-PTZ)(2)(H(2)O)(4)] [4-PTZ is 5-(4-pyridyl N-oxide)tetrazolate, C(6)H(4)N(5)O], (I), in the synthesis of 4-HPTZ, (II), from the cycloaddition reaction of 4-cyanopyridine N-oxide with NaN(3) in water using CdCl(2) as catalyst, was structurally characterized. The unique Cd atom lies on a twofold axis and the coordination geometry of the Cd atom is that of a slightly distorted octahedron, involving four water molecules and two tetrazolate ligands. The catalytic role of the Cd(2+) ion in the tetrazole generation reaction stems from the formation of (I). In acidic solution, (I) can be disassociated and the free ligand, (II), can be released.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
4-Cyanopyridine N-oxide, 96%