Přejít k obsahu
Merck
  • Polymorphisms in GRIK4, HTR2A, and FKBP5 show interactive effects in predicting remission to antidepressant treatment.
Polymorphisms in GRIK4, HTR2A, and FKBP5 show interactive effects in predicting remission to antidepressant treatment.
Neuropsychopharmacology : official publication of the American College of Neuropsychopharmacology (2009-11-20)
Sonja Horstmann, Susanne Lucae, Andreas Menke, Johannes M Hennings, Marcus Ising, Darina Roeske, Bertram Müller-Myhsok, Florian Holsboer, Elisabeth B Binder
ANOTACE
Single-nucleotide polymorphisms (SNPs) in the FKBP5, GRIK4, and HTR2A genes have been shown to be associated with response to citalopram treatment in the STAR(*)D sample, but only associations with FKBP5 have so far been tested in the Munich Antidepressant Response Signature (MARS) project. Response and remission of depressive symptoms after 5 weeks of antidepressant treatment were tested against 82 GRIK4 and 37 HTR2A SNPs. Association analysis was conducted in about 300 depressed patients from the MARS project, 10% of whom had bipolar disorder. The most predictive SNPs from these two genes and rs1360780 in FKBP5 were then genotyped in a total of 387 German depressed in-patients to analyze potential additive and interactive effects of these variants. We could not replicate previous findings of the Sequenced Treatment Alternatives to Relieve Depression (STAR(*)D) study in our sample. Although not statistically significant, the effect for the best GRIK4 SNP of STAR(*)D (rs1954787, p=0.076, p(corrected)=0.98) seemed to be in the same direction. On the other hand, the nominally significant association with the top HTR2A SNPs of STAR(*)D (rs7997012, allelic, p=0.043, p(corrected)=0.62) was with the opposite risk allele. The GRIK4 SNP (rs12800734, genotypic, p=0.0019, p(corrected)=0.12) and the HTR2A SNP (rs17288723, genotypic, p=0.0011, p(corrected)=0.02), which showed the strongest association with remission in our sample, had not been reported previously. Associations across all genetic markers within the GRIK4 (genotypic, p=0.022) or HTR2A (genotypic, p=0.012) locus using the Fisher's product method (FPM) were also significant. In all 374 patients, the best predictive model included a main effect for GRIK4 rs12800734 and two significant interactions between GRIK4 rs12800734 and FKBP5 rs1360780, and GRIK4 rs12800734 and HTR2A rs17288723. This three SNP model explained 13.1% of the variance for remission after 5 weeks (p=0.00051 for the model). Analyzing a sub-sample of 194 patients, plasma ACTH (p=0.002) and cortisol (p=0.021) responses of rs12800734 GG (GRIK4) carriers, who also showed favorable treatment response, were significantly lower in the second combined dexamethasone (dex)/corticotrophin-releasing hormone (CRH) test before discharge compared with the other two genotype groups. Despite large differences in ethnicity and design compared with the STAR(*)D study, our results from the MARS study further support both independent and interactive involvement of GRIK4, HTR2A and FKBP5 in antidepressant treatment response.