- Synthesis, separation, and isomer-dependent packing in two dimensions--detected by scanning tunnelling microscopy--of a TTF derivative.
Synthesis, separation, and isomer-dependent packing in two dimensions--detected by scanning tunnelling microscopy--of a TTF derivative.
Chemical communications (Cambridge, England) (2003-05-13)
Elba Gomar-Nadal, Mohamed M S Abdel-Mottaleb, Steven De Feyter, Jaume Veciana, Concepció Rovira, David B Amabilino, Frans C De Schryver
PMID12739671
ANOTACE
The synthesis, isolation and STM imaging on graphite of the cis and trans isomers of a TTF reveal isomer-dependent packing, and constitutes a way to study the non-covalent interactions at play in these systems.
MATERIÁLY