Přejít k obsahu
Merck
Elevated blood harmane (1-methyl-9H-pyrido[3,4-b]indole) concentrations in essential tremor.
Neurotoxicology (2008-02-05)
Elan D Louis, Wendy Jiang, Kathryn M Pellegrino, Eileen Rios, Pam Factor-Litvak, Claire Henchcliffe, Wei Zheng
ANOTACE
Essential tremor (ET) is a widespread late-life neurological disease. Genetic and environmental factors likely play an etiological role. Harmane (1-methyl-9H-pyrido[3,4-b]indole) is a potent tremor-producing neurotoxin. In 2002, we demonstrated elevated blood harmane concentrations in an initial sample of 100 ET cases compared to 100 controls. Between 2002 and 2007, we assembled a new and larger sample of ET cases and controls. We now attempt to replicate our previous findings. Cases and controls were frequency-matched on age, gender, and race. Blood harmane concentrations were quantified by high-performance liquid chromatography. Subjects comprised 150 ET cases and 135 controls (mean age 65.3+/-15.5 vs. 65.5+/-14.2 years, p=0.94). Mean log blood harmane concentration was approximately 50% higher in cases than controls (0.50+/-0.54g(-10)/ml vs. 0.35+/-0.62g(-10)/ml, p=0.038). In a logistic regression analysis, log blood harmane concentration was associated with ET (OR(adjusted) 1.56, 95% CI 1.01-2.42, p=0.04), and odds of ET was 1.90 (95% CI 1.07-3.39, p=0.029) in the highest versus lowest log blood harmane tertile. Log blood harmane was highest in ET cases with familial ET (0.53+/-0.57g(-10)/ml), intermediate in cases with sporadic ET (0.43+/-0.45g(-10)/ml) and lowest in controls (0.35+/-0.62g(-10)/ml) (test for trend, p=0.026). Blood harmane appears to be elevated in ET. The higher concentrations in familial ET suggests that the mechanism may involve genetic factors.
MATERIÁLY
Číslo produktu
Značka
Popis produktu
Sigma-Aldrich
Harmane, 98%